1-(5-Fluoro-2-nitrophenyl)ethanone 97%

Supplier: Ambeed
Warning

A174096-25G A174096-5G A174096-10G A174096-1G
103806-916EA 25 USD
103806-916 103845-214 77082-500 77082-502 77191-510 77301-314
1-(5-Fluoro-2-nitrophenyl)ethanone 97%
1-(5-Fluoro-2-nitrophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₆FNO₃
Boiling Pt: 296.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09835330
CAS Number: 2250-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR