1-(5-Chloro-2-methylphenyl)ethanol 97%

Supplier: Ambeed
Warning

A102128-250MG A102128-1G A102128-5G
103796-310EA 37 USD
103796-310 103822-002 77188-590 77308-876 77308-878
1-(5-Chloro-2-methylphenyl)ethanol 97%
1-(5-Chloro-2-methylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₁ClO
MW: 170.64 g/mol
Boiling Pt: 254.3±25.0 °C (760 mm Hg)
Flash Pt: 107.6 °C
Storage Temperature: Ambient
MDL Number: MFCD11520985
CAS Number: 58966-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR