5'-Chloro-2'-fluoroacetophenone 95%

Supplier: Ambeed
Warning

Synonyms: 1-(5-Chloro-2-fluorophenyl)ethanone

A286327-10G A286327-1G A286327-5G
103813-706EA 29 USD
103813-706 103834-842 76816-452 76837-374 76837-376
5'-Chloro-2'-fluoroacetophenone 95%
5'-Chloro-2'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆ClFO
MW: 172.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094519
CAS Number: 541508-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR