1-(5-Bromopyrimidin-4-yl)ethanone 97%

Supplier: Ambeed
Warning

A310812-1G A310812-250MG
103814-636EA 353 USD
103814-636 76838-598 77872-430
1-(5-Bromopyrimidin-4-yl)ethanone 97%
1-(5-Bromopyrimidin-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD13193452
CAS Number: 1245643-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR