1-(5-Bromopyridin-3-yl)-2,2,2-trifluoroethanone hydrochloride 95%

Supplier: Ambeed
Warning

A129303-100MG A129303-250MG
103801-260EA 122 USD
103801-260 77149-538 77149-540
1-(5-Bromopyridin-3-yl)-2,2,2-trifluoroethanone hydrochloride 95%
1-(5-Bromopyridin-3-yl)-2,2,2-trifluoroethanone hydrochloride

Precaución: for Research Use Only

Formula: C₇H₄BrClF₃NO
MW: 290.46 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29075654
CAS Number: 1883347-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR