2-Acetyl-5-bromopyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(5-Bromopyridin-2-yl)ethanone
A175295-100G
A175295-10G
A175295-250MG
A175295-1G
A175295-25G
A175295-5G
103807-028EA
8
USD
103807-028
103845-332
76985-606
76985-608
76985-610
77082-670
77082-672
77082-674
2-Acetyl-5-bromopyridine 98%
2-Acetyl-5-bromopyridine
Precaución: for Research Use Only
Formula:
C₇H₆BrNO MW: 200.0300 g/mol Boiling Pt: 257.2 °C (760 mm Hg) Flash Pt: 109.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD04974523 CAS Number: 214701-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...