1-(5-Bromo-2-nitrophenyl)ethanone 98%

Supplier: Ambeed
Warning

Synonyms: 5’-Bromo-2’-nitroacetophenone

A104669-1G A104669-5G A104669-25G
103796-754EA 18 USD
103796-754 103822-472 77309-332 77309-334 77570-276
1-(5-Bromo-2-nitrophenyl)ethanone 98%
1-(5-Bromo-2-nitrophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₆BrNO₃
MW: 244.0400 g/mol
Melting Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD10698995
CAS Number: 41877-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR