1-(5-Bromo-2-methylphenyl)ethanone 97%

Supplier: Ambeed
Warning

A133083-10G A133083-250MG A133083-25G A133083-1G A133083-5G
103801-822EA 76 USD
103801-822 77010-022 77010-024 77010-026 77298-604 77298-606
1-(5-Bromo-2-methylphenyl)ethanone 97%
1-(5-Bromo-2-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₉BrO
MW: 213.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22493313
CAS Number: 90326-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR