5'-Bromo-2'-fluoroacetophenone 98%

Supplier: Ambeed
Warning

A126275-100G A126275-10G A126275-1G A126275-25G A126275-500G
103800-786EA 6 USD
103800-786 76782-032 76782-034 76782-036 77312-986 77312-988 77871-571
5'-Bromo-2'-fluoroacetophenone 98%
5'-Bromo-2'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆BrFO
Boiling Pt: 251 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11226880
CAS Number: 198477-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR