4'-Nitroacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: p-Nitroacetophenone, 1-(4-Nitrophenyl)ethanone

A130814-1000G A130814-100G A130814-500G A130814-25G
77285-300EA 57 USD
77285-300 77285-302 77285-304 77298-432
4'-Nitroacetophenone 98%
4'-Nitroacetophenone

Precaución: for Research Use Only

Formula: C₈H₇NO₃
MW: 165.15 g/mol
Boiling Pt: 275
Melting Pt: 75…78 °C
Flash Pt: 149
Storage Temperature: Ambient
MDL Number: MFCD00007355
CAS Number: 100-19-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR