1-(4-Methoxyphenyl)ethanamine 97%

Supplier: Ambeed
Danger

Synonyms: 1-(4-Methoxyphenyl)ethylamine

A435022-1G A435022-5G A435022-10G A435022-250MG
103827-588EA 13 USD
103827-588 103838-654 76842-368 76842-370 77196-328 77196-330
1-(4-Methoxyphenyl)ethanamine 97%
1-(4-Methoxyphenyl)ethanamine

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MW: 151.21 g/mol
Boiling Pt: 240.3 °C (760 mm Hg)
Flash Pt: 99.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00044523
CAS Number: 6298-96-0
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR