1-(4-Methoxyphenyl)ethanol 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxy-alpha-methylbenzyl alcohol

A181385-1G A181385-25G A181385-5G A181385-100G A181385-10G
103807-598EA 6 USD
103807-598 103845-970 77083-498 77083-500 77083-502 77191-684 77191-686
1-(4-Methoxyphenyl)ethanol 97%
1-(4-Methoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 254.3 °C (760 mm Hg)
Flash Pt: 73.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00016857
CAS Number: 3319-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR