Benzyl-4-methoxyphenyl ketone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4'-Methoxy-2-phenylacetophenone, 1-(4-Methoxyphenyl)-2-phenylethanone
A180402-1G
A180402-25G
A180402-5G
A180402-10G
A180402-250MG
103807-536EA
12
USD
103807-536
103845-892
77083-380
77083-382
77083-384
77260-200
77260-202
Benzyl-4-methoxyphenyl ketone 98%
Benzyl 4-methoxyphenyl ketone
Precaución: for Research Use Only
Formula:
C₁₅H₁₄O₂ MW: 226.2700 g/mol Boiling Pt: 380 °C (760 mm Hg) Melting Pt: 71…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017177 CAS Number: 1023-17-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...