1-(4-Methoxynaphthalen-1-yl)ethanone 97%

Supplier: Ambeed
Warning

A103346-100G A103346-250MG A103346-1G A103346-5G A103346-10G
103796-532EA 7 USD
103796-532 103822-238 77188-652 77188-654 77283-078 77283-080 77745-786
1-(4-Methoxynaphthalen-1-yl)ethanone 97%
1-(4-Methoxynaphthalen-1-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₃H₁₂O₂
MW: 200.23 g/mol
Boiling Pt: 366.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00156696
CAS Number: 24764-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR