N-(4-Methoxybenzyl)urea 97%

Supplier: Ambeed
Warning

A118936-10G A118936-1G A118936-5G A118936-25G A118936-100G
103799-136EA 6 USD
103799-136 76995-682 77297-646 77297-648 77670-676 77880-964
N-(4-Methoxybenzyl)urea 97%
N-(4-Methoxybenzyl)urea

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₂
Boiling Pt: 327.9 °C (760 mm Hg)
Flash Pt: 152.1 °C
Storage Temperature: Ambient
MDL Number: MFCD01314623
CAS Number: 54582-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR