1-(4-Fluorophenyl)-2-pyrrolidinone 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(4-Fluorophenyl)pyrrolidin-2-one
A199999-250MG
A199999-1G
103817-154EA
50
USD
103817-154
103847-780
77192-250
77192-252
77302-642
1-(4-Fluorophenyl)-2-pyrrolidinone 95%
1-(4-Fluorophenyl)-2-pyrrolidinone
Precaución: for Research Use Only
Formula:
C₁₀H₁₀FNO Boiling Pt: 375.1 °C (760 mm Hg) Flash Pt: 180.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00138504 CAS Number: 54660-08-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...