4'-Fluoropropiophenone 98%

Supplier: Ambeed
Warning

A103443-100G A103443-10G A103443-25G A103443-500G A103443-5G A103443-1000G
103822-256EA 6 USD
103822-256 77283-092 77283-094 77283-096 77283-098 77309-126 77747-770
4'-Fluoropropiophenone 98%
4'-Fluoropropiophenone

Precaución: for Research Use Only

Formula: C₉H₉FO
Boiling Pt: 215.9 °C (760 mm Hg)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00000356
CAS Number: 456-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR