1-(4-Fluorophenyl)cyclopropanecarboxylic acid 96%

Supplier: Ambeed
Warning

A267603-100G A267603-100MG A267603-10G A267603-250MG A267603-1G A267603-25G A267603-5G
103812-866EA 18 USD
103812-866 103852-766 76988-270 76988-272 76988-274 76988-276 77092-512 77092-514 77304-982
1-(4-Fluorophenyl)cyclopropanecarboxylic acid 96%
1-(4-Fluorophenyl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉FO₂
MW: 180.1800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06208777
CAS Number: 773100-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR