4-Chlorophenylurea 98%

Supplier: Ambeed
Danger

Synonyms: 1-(4-Chlorophenyl)urea

A697335-5G A697335-10G A697335-1G
103840-614EA 30 USD
103840-614 76844-836 77293-254 77308-142
4-Chlorophenylurea 98%
4-Chlorophenylurea

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O
MW: 170.6 g/mol
Boiling Pt: 271.7 °C (760 mm Hg)
Melting Pt: 204…205 °C
Flash Pt: 118.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00014788
CAS Number: 140-38-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR