1-(4-Chlorophenyl)-N-methylmethanamine 95%

Supplier: Ambeed
Danger

Synonyms: 4-Chloro-N-methylbenzylamine

A244459-1G A244459-25G A244459-10G A244459-5G
103811-582EA 14 USD
103811-582 103851-414 77090-622 77090-624 77113-308 77304-320
1-(4-Chlorophenyl)-N-methylmethanamine 95%
1-(4-Chlorophenyl)-N-methylmethanamine

Precaución: for Research Use Only

Formula: C₈H₁₀ClN
MW: 155.6247 g/mol
Flash Pt: 78.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00018749
CAS Number: 104-11-0
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR