1-(4-Chlorophenyl)cyclopropanecarboxylic acid 95%

Supplier: Ambeed
Warning

A810058-10G A810058-25G A810058-5G A810058-1G
103841-088EA 42 USD
103841-088 76807-570 76845-340 76845-342 77308-366
1-(4-Chlorophenyl)cyclopropanecarboxylic acid 95%
1-(4-Chlorophenyl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉ClO₂
MW: 196.64 g/mol
Boiling Pt: 336.7 °C (760 mm Hg)
Melting Pt: 152…155 °C
Flash Pt: 157.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00001289
CAS Number: 72934-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR