1-(4-Chlorophenyl)-2-methylpropan-1-one 97%

Supplier: Ambeed
Warning

A197145-100MG A197145-250MG
103816-898EA 119 USD
103816-898 103847-516 77900-663 77900-843
1-(4-Chlorophenyl)-2-methylpropan-1-one 97%
1-(4-Chlorophenyl)-2-methylpropan-1-one

Precaución: Sealed in dry, at room temperature.

Formula: C₁₀H₁₁ClO
Boiling Pt: 264.1 °C (760 mm Hg)
Flash Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD01696607
CAS Number: 18713-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR