4'-Chloro-3'-methylacetophenone 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(4-Chloro-3-methylphenyl)ethanone
A130087-1G
A130087-25G
103801-376EA
11
USD
103801-376
77313-808
77313-810
4'-Chloro-3'-methylacetophenone 95%
4'-Chloro-3'-methylacetophenone
Precaución: for Research Use Only
Formula:
C₉H₉ClO MW: 168.62 g/mol Boiling Pt: 263 °C (760 mm Hg) Flash Pt: 126.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00045172 CAS Number: 37074-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...