4'-Chloro-2'-nitroacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Chloro-2-nitrophenyl)ethanone

A132829-100MG A132829-250MG A132829-1G A132829-5G
103801-792EA 47 USD
103801-792 76870-824 76870-826 77314-376 77314-378
4'-Chloro-2'-nitroacetophenone 98%
4'-Chloro-2'-nitroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆ClNO₃
MW: 199.5900 g/mol
Melting Pt: 55…56 °C
Storage Temperature: Ambient
MDL Number: MFCD09031331
CAS Number: 23082-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR