1-(4-Chloro-2-(3-methyl-1H-pyrazol-1-yl)phenyl)-2,2,2-trifluoroethanone 97%

Supplier: Ambeed
Warning

A175567-250MG A175567-1G
103807-060EA 19 USD
103807-060 103845-372 77011-744 77082-712
1-(4-Chloro-2-(3-methyl-1H-pyrazol-1-yl)phenyl)-2,2,2-trifluoroethanone 97%
1-(4-Chloro-2-(3-methyl-1H-pyrazol-1-yl)phenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₁₂H₈ClF₃N₂O
MW: 288.65 g/mol
Boiling Pt: 365.4±42.0 °C (760 mm Hg)
Flash Pt: 174.8 °C
Storage Temperature: Ambient
MDL Number: MFCD14706009
CAS Number: 1125828-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR