2-Acetyl-4-bromopyridine 97%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Bromo-pyridin-2-yl)-ethanone, 1-(4-Bromopyridin-2-yl)ethanone

A871093-100MG A871093-250MG A871093-1G
103841-284EA 444 USD
103841-284 76832-668 76832-670 76845-540
2-Acetyl-4-bromopyridine 97%
2-Acetyl-4-bromopyridine

Precaución: for Research Use Only

Formula: C₇H₆BrNO
MW: 200.035 g/mol
Boiling Pt: 267.3±25.0 °C (760 mm Hg)
Flash Pt: 115.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13189212
CAS Number: 1060805-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR