1-(4-Bromophenyl)imidazole 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Bromophenyl)-1H-imidazole

A832029-250MG A832029-1G A832029-5G
103841-170EA 143 USD
103841-170 76698-258 77308-416 77308-418
1-(4-Bromophenyl)imidazole 98%
1-(4-Bromophenyl)imidazole

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MW: 223.07 g/mol
Boiling Pt: 323.4 °C (760 mm Hg)
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00060489
CAS Number: 10040-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR