1-(4-Bromophenyl)ethane-1,2-diol 98%
Supplier: Ambeed
|
Warning
|
A176491-100MG
A176491-250MG
A176491-1G
103807-142EA
301
USD
103807-142
103845-470
76872-586
76872-588
77082-822
1-(4-Bromophenyl)ethane-1,2-diol 98%
1-(4-Bromophenyl)ethane-1,2-diol
Precaución: for Research Use Only
Formula:
C₈H₉BrO₂ Storage Temperature: Ambient |
MDL Number:
MFCD09743652 CAS Number: 92093-23-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...