1-(4-Bromonaphthalen-1-yl)ethanone 95%

Supplier: Ambeed
Warning

A155043-250MG A155043-1G
103804-708EA 51 USD
103804-708 103842-904 77162-934 77318-638
1-(4-Bromonaphthalen-1-yl)ethanone 95%
1-(4-Bromonaphthalen-1-yl)ethanone

Precaución: Sealed in dry, at room temperature.

Formula: C₁₂H₉BrO
MW: 249.1 g/mol
Flash Pt: 114 °C
Storage Temperature: Ambient
MDL Number: MFCD09037858
CAS Number: 46258-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR