1-(4-Bromo-2-fluorophenyl)propan-1-one 95%

Supplier: Ambeed
Warning

A153511-10G A153511-250MG A153511-1G A153511-5G
103804-522EA 44 USD
103804-522 103842-696 76871-740 76871-742 77318-358 77318-360
1-(4-Bromo-2-fluorophenyl)propan-1-one 95%
1-(4-Bromo-2-fluorophenyl)propan-1-one

Precaución: for Research Use Only

Formula: C₉H₈BrFO
MW: 231.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226830
CAS Number: 259750-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR