4'-Bromo-2'-fluoroacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Bromo-2-fluorophenyl)ethanone

A121088-250MG A121088-25G A121088-1G A121088-5G A121088-10G A121088-100G
103800-028EA 6 USD
103800-028 77234-114 77234-116 77312-144 77312-146 77571-262 77745-891
4'-Bromo-2'-fluoroacetophenone 97%
4'-Bromo-2'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆BrFO
MW: 217.04 g/mol
Flash Pt: 108.81 °C
Storage Temperature: Ambient
MDL Number: MFCD03411551
CAS Number: 625446-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR