4'-Bromo-2',6'-difluoroacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Bromo-2,6-difluorophenyl)ethanone, 1-(4-Bromo-2,6-difluorophenyl)ethan-1-one

A267668-100MG A267668-10G A267668-250MG A267668-1G
103812-872EA 20 USD
103812-872 103852-772 76988-278 76988-280 76988-282 77092-518
4'-Bromo-2',6'-difluoroacetophenone 97%
4'-Bromo-2',6'-difluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O
MW: 235.026 g/mol
Boiling Pt: 246.1±40.0 °C
Flash Pt: 102.6 °C
Storage Temperature: Ambient
MDL Number: MFCD22570410
CAS Number: 746630-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR