4-Bromo-2,6-difluoro-α-methylbenzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2,6-difluoro-.alpha.-methyl-benzenemethanol, 1-(4-Bromo-2,6-difluorophenyl)ethanol

A218117-1G A218117-5G
103818-694EA 24 USD
103818-694 103849-422 77088-076 77088-078
4-Bromo-2,6-difluoro-α-methylbenzyl alcohol 97%
4-Bromo-2,6-difluoro-α-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O
Boiling Pt: 249.1 °C (760 mm Hg)
Flash Pt: 104.5 °C
Storage Temperature: Ambient
MDL Number: MFCD25459281
CAS Number: 1214900-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR