1-Acetylpiperidin-4-amine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 1-Acetyl-4-aminopiperidine hydrochloride, 1-(4-Aminopiperidin-1-yl)ethanone hydrochloride

A188396-1G A188396-25G A188396-5G A188396-100MG A188396-10G A188396-250MG
103816-094EA 19 USD
103816-094 103846-690 77084-418 77084-420 77084-422 77191-874 77191-876 77191-878
1-Acetylpiperidin-4-amine hydrochloride 95%
1-Acetylpiperidin-4-amine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₅ClN₂O
MW: 178.6600 g/mol
Melting Pt: 240…245 °C
Storage Temperature: Ambient
MDL Number: MFCD08690151
CAS Number: 214147-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR