1-(4-Aminophenyl)-3-morpholino-5,6-dihydropyridin-2(1H)-one 95%

Supplier: Ambeed
Warning

A431106-5G A431106-100G A431106-10G
103827-520EA 24 USD
103827-520 103838-586 76842-280 77168-652 77168-654
1-(4-Aminophenyl)-3-morpholino-5,6-dihydropyridin-2(1H)-one 95%
1-(4-Aminophenyl)-3-morpholino-5,6-dihydropyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₅H₁₉N₃O₂
MW: 273.33 g/mol
Boiling Pt: 491.5±45.0 °C (760 mm Hg)
Melting Pt: 195…199 °C
Flash Pt: 251 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22581648
CAS Number: 1267610-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR