Agmatine sulfate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Amino-4-guanidobutane Sulphate, 1-(4-Aminobutyl)guanidine sulphate

A177688-100G A177688-500G A177688-25G A177688-5G
103845-592EA 14 USD
103845-592 76690-422 76690-424 77301-518 77301-520
Agmatine sulfate 98%
Agmatine sulfate

Precaución: for Research Use Only

Formula: C₅H₁₄N₄
Boiling Pt: 281.4 °C (760 mm Hg)
Melting Pt: 234…238 °C
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD00013109
CAS Number: 2482-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR