1-(4-Aminobutyl)-1H-pyrrole-2,5-dione hydrochloride 97%

Supplier: Ambeed
Warning

A503312-100MG A503312-250MG
103828-312EA 223 USD
103828-312 77144-042 77144-044
1-(4-Aminobutyl)-1H-pyrrole-2,5-dione hydrochloride 97%
1-(4-Aminobutyl)-1H-pyrrole-2,5-dione hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₃ClN₂O₂
MW: 204.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16619569
CAS Number: 952292-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR