4'-Amino-2'-hydroxyacetophenone 96%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Amino-2-hydroxyphenyl)ethanone, 1-(4-Amino-2-hydroxyphenyl)ethan-1-one

A193469-1G A193469-25G A193469-5G A193469-100MG A193469-250MG
103816-540EA 11 USD
103816-540 103847-156 77085-060 77085-062 77085-064 77192-048 77192-050
4'-Amino-2'-hydroxyacetophenone 96%
4'-Amino-2'-hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.1600 g/mol
Boiling Pt: 331.7 °C (760 mm Hg)
Melting Pt: 134…135 °C
Flash Pt: 154.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00100636
CAS Number: 2476-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR