1-(4,6-Dichloropyrimidin-5-yl)ethanone 97%

Supplier: Ambeed
Warning

A121816-100MG A121816-250MG A121816-25G A121816-1G A121816-5G
103800-138EA 23 USD
103800-138 77009-454 77009-456 77009-458 77312-268 77312-270
1-(4,6-Dichloropyrimidin-5-yl)ethanone 97%
1-(4,6-Dichloropyrimidin-5-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD13689148
CAS Number: 60025-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR