4'-(Methylthio)acetophenone 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(4-(Methylthio)phenyl)ethanone
A306602-25G
A306602-5G
A306602-100G
A306602-10G
103835-648EA
13
USD
103835-648
76838-374
76838-376
77002-594
77002-596
4'-(Methylthio)acetophenone 97%
4'-(Methylthio)acetophenone
Precaución: for Research Use Only
Formula:
C₉H₁₀OS MW: 166.24 g/mol Boiling Pt: 135 °C (1 mm Hg) Melting Pt: 80…82 °C Flash Pt: 86.848 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039835 CAS Number: 1778-09-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...