4-Acetyl-N-methylaniline 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-(Methylamino)phenyl)ethanone, 4'-(Methylamino)acetophenone

A210917-100MG A210917-250MG A210917-1G A210917-5G A210917-25G
103818-054EA 23 USD
103818-054 103848-746 76986-712 76986-714 77087-132 77087-134 77303-158
4-Acetyl-N-methylaniline 98%
4-Acetyl-N-methylaniline

Precaución: for Research Use Only

Formula: C₉H₁₁NO
MW: 149.193 g/mol
Boiling Pt: 281.7±23.0 °C (760 mm Hg)
Melting Pt: 103…107 °C
Flash Pt: 125.8 °C
Storage Temperature: Ambient
MDL Number: MFCD09261160
CAS Number: 17687-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR