4'-(Dimethylamino)acetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-(Dimethylamino)phenyl)ethanone

A231361-100G A231361-10G A231361-1G A231361-500G A231361-25G A231361-5G
103850-522EA 6 USD
103850-522 76771-162 76771-164 76771-166 76771-168 77089-410 77089-412
4'-(Dimethylamino)acetophenone 98%
4'-(Dimethylamino)acetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO
MW: 163.2200 g/mol
Boiling Pt: 282.9 °C (760 mm Hg)
Melting Pt: 105 °C
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00017246
CAS Number: 2124-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR