1-[4-(Chloromethyl)phenyl]-1H-pyrazole 97%

Supplier: Ambeed
Danger

Synonyms: 1-(4-(Chloromethyl)phenyl)-1H-pyrazole

A315290-1G A315290-250MG A315290-25G
103825-060EA 20 USD
103825-060 103835-986 76838-822 77099-766 77306-126
1-[4-(Chloromethyl)phenyl]-1H-pyrazole 97%
1-[4-(Chloromethyl)phenyl]-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂
MW: 192.645 g/mol
Boiling Pt: 303.9 °C (760 mm Hg)
Flash Pt: 137.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07366379
CAS Number: 143426-52-2
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR