1-(4-(Bromomethyl)phenyl)ethanone 97%

Supplier: Ambeed
Danger

A132828-100MG A132828-250MG A132828-5G A132828-1G A132828-25G
103801-790EA 31 USD
103801-790 76782-438 76782-440 77298-586 77314-372 77314-374
1-(4-(Bromomethyl)phenyl)ethanone 97%
1-(4-(Bromomethyl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₉BrO
Boiling Pt: 291.4 °C (760 mm Hg)
Melting Pt: 38.5…40.0 °C
Flash Pt: 94.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10001463
CAS Number: 51229-51-7
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR