1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropanecarbonitrile 97%

Supplier: Ambeed
Warning

A162608-100MG A162608-10G A162608-250MG A162608-1G A162608-5G
103805-638EA 58 USD
103805-638 103843-888 76812-838 76812-840 76812-842 77319-946 77319-948
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropanecarbonitrile 97%
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropanecarbonitrile

Precaución: for Research Use Only

Formula: C₁₆H₂₀BNO₂
MW: 269.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18383917
CAS Number: 1206641-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR