1-Acetyl-5,6-dihydro-2H-pyridine-4-boronic acid, pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridin-1(2H)-yl)ethanone

A131034-100MG A131034-250MG A131034-1G A131034-5G
103801-514EA 21 USD
103801-514 76870-736 76870-738 77313-990 77313-992
1-Acetyl-5,6-dihydro-2H-pyridine-4-boronic acid, pinacol ester 97%
1-Acetyl-5,6-dihydro-2H-pyridine-4-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₂BNO₃
MW: 251.13 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18427628
CAS Number: 1227068-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR