1-(3-Pyridyl)-3-(dimethylamino)-2-propen-1-one 96%

Supplier: Ambeed
Warning

Synonyms: 3-(Dimethylamino)-1-(pyridin-3-yl)prop-2-en-1-one

A111729-100G A111729-10G A111729-25G A111729-500G A111729-5G A111729-1000G
103808-070EA 502 USD
103808-070 77108-004 77108-006 77108-008 77108-010 77108-012 77284-178
1-(3-Pyridyl)-3-(dimethylamino)-2-propen-1-one 96%
1-(3-Pyridyl)-3-(dimethylamino)-2-propen-1-one

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O
MW: 176.22 g/mol
Boiling Pt: 281.4 °C (760 mm Hg)
Melting Pt: 86…88 °C
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD00115180
CAS Number: 55314-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR