1-(3-Methoxypyridin-2-yl)ethanone 97%
Supplier: Ambeed
|
Warning
|
A231885-100MG
A231885-250MG
A231885-1G
A231885-5G
103810-800EA
48
USD
103810-800
103850-554
76826-470
76826-472
77089-456
77089-458
1-(3-Methoxypyridin-2-yl)ethanone 97%
1-(3-Methoxypyridin-2-yl)ethanone
Precaución: for Research Use Only
Formula:
C₈H₉NO₂ Boiling Pt: 238.8±20.0 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD18256469 CAS Number: 379227-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...