(3-Methoxybenzyl)methylamine 97%

Supplier: Ambeed
Danger

Synonyms: 3-Methoxy-N-methylbenzylamine, 1-(3-Methoxyphenyl)-N-methylmethanamine

A690507-1G A690507-10G A690507-25G A690507-5G
103840-578EA 63 USD
103840-578 76844-794 77280-760 77308-126 77308-128
(3-Methoxybenzyl)methylamine 97%
(3-Methoxybenzyl)methylamine

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MW: 151.21 g/mol
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD04618244
CAS Number: 41789-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR