1-(3-Hydroxypyrrolidin-1-yl)ethanone 95%

Supplier: Ambeed
Warning

A102976-250MG A102976-5G A102976-100MG
103796-458EA 62 USD
103796-458 103822-162 76821-314 77309-030 77884-931
1-(3-Hydroxypyrrolidin-1-yl)ethanone 95%
1-(3-Hydroxypyrrolidin-1-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD09909312
CAS Number: 23123-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR